Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements

Catalog No. T001011000 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
25 L
Packaging:
Aluminum Jug

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Acetonitrile is suitable for HPLC, GC, SFC, TLC, ACS, Spectrophotometry, and meets the analytical requirements of USP. | CAS: 75-05-8 | C2H3N | 41.05 g/mol

Chromplete Acetonitrile meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -51°F
Absorbance <0.1 at 195 nm, <0.05 at 200 nm, <0.02 at 210 nm, <0.01 at 220 nm
Color ≤ 5 APHA
CAS Min % 0.9995
Density 0.775 - 0.78
Boiling Point 180°F
Quantity 1 L
Packaging Aluminum Jug
Physical Form Liquid
Refractive Index 1.3405 – 1.3425
Residue after Evaporation ≤ 0.8 ppm
Titratable Acid ≤ 0.008 mEq/g
Titratable Base ≤ 0.0006 mEq/g
UN Number 1648
Water ≤ 0.01 %
Formula Weight 41.05
Percent Purity 0.9995
Grade Chromplete
Assay Minimum, GC-FID
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Flammable liquids (category 2), Acute toxicity (category 4)
Hazard Statement Causes serious eye irritation, Highly flammable liquid and vapor, Harmful if swallowed, in contact with skin or if inhaled
ShelfLife 1825 days
EINECSNumber 200-835-2
RTECSNumber AL7700000
storageNote1 In sealed containers
Recommended Storage Room temperature flammable storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.