Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromplete™ Water, HPLC/GC, and Spectrometry
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Description
Chromplete Water is suitable for use in:
- HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
- GC- FID, GC- ECD, GC-TCD
- UV Spectrophotometry
- Preparative chromatography
- Supercritical Fluid and Thin Layer chromatography
- Reactions/Analyses requiring ACS compliance
- Extraction/Concentration Analyses
Specifications
Specifications
| Absorbance | ≤ 0.01, ≤ 0.005 |
| pH | 5.5-7.0 |
| Color | ≤5 APHA |
| Melting Point | 0°C |
| CAS Min % | 0.99 |
| Boiling Point | 100°C |
| Formula Weight | 18.01 |
| Grade | Chromplete |
| Packaging | Amber Glass |
| Residue after Evaporation | ≤ 1.0 ppm |
| Show More |
Safety and Handling
| ShelfLife | 1825 Days |
| EINECSNumber | 231-791-2 |
| RTECSNumber | ZC0110000 |
| storageNote1 | At room temperature in sealed containers |
| Recommended Storage | Room Temperature |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction