Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrysin, 99+%

Catalog No. ac110320050
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 480-40-0 | C15H10O4 | 254.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-40-0
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00006834
InChI Key RTIXKCRFFJGDFG-UHFFFAOYSA-N
Synonym chrysin, 5,7-dihydroxyflavone, chrysine, 5,7-dihydroxy-2-phenyl-4h-chromen-4-one, crysin, 4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl, unii-3cn01f5zj5, flavone, 5,7-dihydroxy, 5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one, chembl117
PubChem CID 5281607
ChEBI CHEBI:75095
IUPAC Name 5,7-dihydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O

Specifications

Melting Point 284.0°C to 290.0°C
Color Yellow
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 18,124
Merck Index 15,226
Formula Weight 254.24
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Chrysin
Show More Show Less
EINECSNumber 207-549-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.