Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CHS 828
GREENER_CHOICESDP

Catalog No. 67535 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-535 5 mg
67-532-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-535 Supplier Tocris Bioscience™ Supplier No. 6753/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

NAMPT inhibitor; active in vivo and cytotoxic

CHS 828 is a NAMPT inhibitor. Decreases cellular level of NAD+. Selectively increases intracellular ROS levels in cancer cells but not normal cells. Active in vivo and cytotoxic.

Specifications

Chemical Name or Material N-[6-(4-Chlorophenoxy)hexyl]-N'-cyano-N''-4-pyridinylguanidine
CAS 200484-11-3
Quantity 5 mg
Molecular Formula C19H22ClN5O
Formula Weight 371.86
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.