Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572664
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572664 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572664 Supplier Invitrogen™ Supplier No. PA572664
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHST1 Polyclonal Antibody for Western Blot

Carbohydrate sulfotransferase 1 (CHST1), also referred to as KSGal6ST or KSST, is a keratan sulfate sulfotransferase. It is predominantly expressed in brain and skeletal muscle and localizes to the trans-Golgi network. CHST1 is responsible for mediating the sulfation of keratan in the cornea, which is important in maintaining corneal transparency. In particular, CHST1 catalyzes the transfer of sulfate groups from 3'-phosphoadenosine 5'-phosphosulfate to position six of internal or terminal galactose residues (preferentially the residues adjacent to sulfated GlcNAc) on growing keratan sulfate chains. CHST1 also contributes to the generation of L-selectin ligands. Mutations in the CHST1 gene may play a role in macular corneal dystrophy (MCD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST1
Applications Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CHST1
Gene Accession No. Q6DBY9
Gene Alias 2610008E20Rik; AW125896; C6ST; carbohydrate (chondroitin 6/keratan) sulfotransferase 1; carbohydrate (keratan sulfate Gal-6) sulfotransferase 1; carbohydrate sulfotransferase 1; chst1; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 1; Gst1; GST-1; hypothetical protein LOC445124; keratan sulfate Gal-6 sulfotransferase; Keratan sulfotransferase; KS6ST; KSGal6ST; KSST; N-acetylglucosamine-6-O-sulfotransferase 1; sulfo1; sulfotransferase1; zgc:100904; zgc:100904 protein
Gene Symbols CHST1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 341-375 amino acids of DANRE chst1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445124
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.