Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546786 Supplier Invitrogen™ Supplier No. PA546786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YRHEIAPGII RKYRRNRTET RGIQFEDFVR YLGDPNHRWL DLQFGDHIIH.

This protein encoded by this gene transfers sulfate to the C-3 hydroxyl of terminal glucuronic acid of protein- and lipid-linked oligosaccharides. This protein was first identified as a sulfotransferase that acts on the human natural killer-1 (HNK-1) glycan; HNK-1 is a carbohydrate involved in neurodevelopment and synaptic plasticity.[provided by RefSeq, Feb 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CHST10
Gene Accession No. O43529
Gene Alias AI507003; AU041319; carbohydrate sulfotransferase 10; CHST10; HNK-1 sulfotransferase; HNK1ST; HNK-1ST; huHNK-1ST; raHNK-1ST; ST; sul-T
Gene Symbols CHST10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CHST10 (aa 213-262).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9486
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.