Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568129 Supplier Invitrogen™ Supplier No. PA568129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of chondroitin. Chondroitin sulfate constitutes the predominant proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices. Can also sulfate Gal residues in desulfated dermatan sulfate. Preferentially sulfates in GlcA->GalNAc unit than in IdoA->GalNAc unit. Does not form 4, 6- di-O-sulfated GalNAc when chondroitin sulfate C is used as an acceptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST11
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHST11
Gene Accession No. P69478, Q9JME2, Q9NPF2
Gene Alias 1110020P09Rik; C4s; C4S-1; C4ST; C4ST1; C4ST-1; carbohydrate (chondroitin 4) sulfotransferase 11; carbohydrate sulfotransferase 11; chondroitin 4; chondroitin 4-O-sulfotransferase 1; Chondroitin 4-sulfotransferase 1; chondroitin 4-sulfotransferase-1; CHST11; HSA269537; IgH/CHST11 fusion; LOW QUALITY PROTEIN: carbohydrate sulfotransferase 11; zC4ST-1
Gene Symbols CHST11
Host Species Rabbit
Immunogen Synthesized peptide derived from human CHST11 (aa 207-230).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314694, 50515, 58250
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.