Learn More
Invitrogen™ CHST12 Monoclonal Antibody (3D6)

Description
Peptide Sequence: RDRELTADSD VDEFLDKFLS AGVKQSDLPR KETEQPPAPG SMEESVRGYD WSPRDARRSP DQGRQQAERR SVLRGFCANS SLAFPTKERA FDDIP.
Specifications
Specifications
| Antigen | CHST12 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3D6 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Chst12 |
| Gene Accession No. | Q9NRB3 |
| Gene Alias | AI595374; C4S-2; C4ST2; C4ST-2; carbohydrate (chondroitin 4) sulfotransferase 12; carbohydrate sulfotransferase 12; Chondroitin 4-O-sulfotransferase 2; chondroitin 4-sulfotransferase 2; Chst12; Sulfotransferase Hlo; UNQ500/PRO1017 |
| Gene Symbols | Chst12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.