Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559672 Supplier Invitrogen™ Supplier No. PA559672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGKLETLDED AAQLLQLLQV DRQLRFPPSY RNRTASSWEE DWFAKIPLAW RQQLYKLYEA DFVLFGYPKP ENLLRD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of chondroitin and desulfated dermatan sulfate. Chondroitin sulfate constitutes the predominant proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices. Activity toward partially desulfated dermatan sulfate is however lower. Does not form 4, 6-di-O-sulfated GalNAc when chondroitin sulfate C is used as an acceptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Chst12
Gene Accession No. Q9NRB3
Gene Alias AI595374; C4S-2; C4ST2; C4ST-2; carbohydrate (chondroitin 4) sulfotransferase 12; carbohydrate sulfotransferase 12; Chondroitin 4-O-sulfotransferase 2; chondroitin 4-sulfotransferase 2; Chst12; Sulfotransferase Hlo; UNQ500/PRO1017
Gene Symbols Chst12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHST12. Recombinant protein control fragment (Product #RP-98447).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55501
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.