Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHST12 Polyclonal Antibody
CHST12 Polyclonal Antibody
Supplier: Thermo Scientific PA5103699
Description
CHST12 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
CHST12 | |
Polyclonal | |
Unconjugated | |
Chst12 | |
AI595374; C4S-2; C4ST2; C4ST-2; carbohydrate (chondroitin 4) sulfotransferase 12; carbohydrate sulfotransferase 12; Chondroitin 4-O-sulfotransferase 2; chondroitin 4-sulfotransferase 2; Chst12; Sulfotransferase Hlo; UNQ500/PRO1017 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
304322, 55501, 59031 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q99LL3, Q9NRB3 | |
Chst12 | |
A synthesized peptide derived from human CHST12(Accession Q9NRB3), corresponding to amino acid residues E343-Y393. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction