Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113888 Supplier Invitrogen™ Supplier No. PA5113888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CHPCRLRYDV VGKFETLAED AAFVLGLAGA SDLSFPGPPR PRGAAASRDL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Guinea Pig: 77%; Human: 100%.

The protein encoded by this gene belongs to the sulfotransferase 2 family. It is localized to the Golgi membrane, and catalyzes the transfer of sulfate to the C4 hydroxyl of beta-1, 4-linked N-acetylgalactosamine (GalNAc) flanked by glucuronic acid residue in chondroitin. Chondroitin sulfate constitutes the predominant proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST13
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CHST13
Gene Accession No. Q8NET6
Gene Alias 1110067M19Rik; C4ST3; C4ST-3; carbohydrate (chondroitin 4) sulfotransferase 13; carbohydrate sulfotransferase 13; Chondroitin 4-O-sulfotransferase 3; chondroitin 4-sulfotransferase 3; CHST13; DcR3; Decoy receptor 3; M68; RGD1562825
Gene Symbols CHST13
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human CHST13 (aa 254-303).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166012
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.