Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566101 Supplier Invitrogen™ Supplier No. PA566101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AYRNKFGEIRE YQQRYGAEIV RRYRAGAGPS PAGDDVTFPE FLRYLVDEDP ERMNEHWMPV YHLCQPCAVH Y Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of dermatan sulfate. Plays a pivotal role in the formation of 4-0-sulfated IdoA blocks in dermatan sulfate. Transfers sulfate to the C-4 hydroxyl of beta1,4-linked GalNAc that is substituted with an alpha-linked iduronic acid (IdoUA) at the C-3 hydroxyl. Transfers sulfate more efficiently to GalNAc residues in -IdoUA-GalNAc-IdoUA- than in -GlcUA-GalNAc-GlcUA-sequences. Has preference for partially desulfated dermatan sulfate. Addition of sulfate to GalNAc may occur immediately after epimerization of GlcUA to IdoUA. GlcUA to IdoUA. Appears to have an important role in the formation of the cerbellar neural network during postnatal brain development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST14
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHST14
Gene Accession No. Q8NCH0
Gene Alias 2600016L03Rik; ATCS; carbohydrate (dermatan 4) sulfotransferase 14; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14; carbohydrate sulfotransferase 14; CHST14; D4ST1; D4ST-1; dermatan 4 sulfotransferase 1; dermatan 4-sulfotransferase 1; dermatan-4-sulfotransferase-1; EDSMC1; hD4ST1; HNK1ST; UNQ1925/PRO4400
Gene Symbols CHST14
Host Species Rabbit
Immunogen Recombinant Human CHST14. Recombinant protein control fragment (Product #RP-106795).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.