Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597171 Supplier Invitrogen™ Supplier No. PA597171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQVGLYAVYL LDWLSVFDKQ QFLILRLEDH ASNVKYTMHK VFQFLNLGPL SEKQEALMTK SPASNARRPE DRNLGPMWPI TQKILRDFYR PFNARLAQVL ADEAFAWKTT; Positive Samples: Raji, Rat heart; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

GALNAC4S-6ST is a sulfotransferase that transfers sulfate from 3'-phosphoadenosine 5'-phosphosulfate (PAPS) to the C-6 hydroxyl group of the GalNAc 4-sulfate residue of chondroitin sulfate A and forms chondroitin sulfate E containing GlcA-GalNAc(4, 6-SO(4)) repeating units. It also transfers sulfate to a unique non-reducing terminal sequence, GalNAc(4SO4)-GlcA(2SO4)-GalNAc(6SO4), to yield a highly sulfated structure similar to the structure found in thrombomodulin chondroitin sulfate. GALNAC4S-6ST may also act as a B-cell receptor involved in BCR ligation-mediated early activation that mediate regulatory signals key to B-cell development and/or regulation of B-cell-specific RAG expression; however such results are unclear in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CHST15
Gene Accession No. Q7LFX5, Q8CHI9
Gene Alias 4631426J05Rik; B cell RAG associated protein; B cell RAG associated protein (GALNAC4S-6ST); B-cell RAG-associated gene protein; Brag; carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15; carbohydrate sulfotransferase 15; Chst15; DKFZp781H1369; GalNAc 4-sulfate 6-O-sulfotransferase; GALNAC4S6ST; GALNAC4S-6ST; GalNAcS-6ST; hBRAG; KIAA0598; MAd5; MGC34346; mKIAA0598; N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase; RP11-47G11.1
Gene Symbols CHST15
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 452-561 of human CHST15 (NP_0569762).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286974, 51363
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.