Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564950 Supplier Invitrogen™ Supplier No. PA564950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HALGAMEVICN SMAKTLQTAL QPPDWLQGHY LVVRYEDLVG DPVKTLRRVY DFVGLLVSPE MEQFALNMTS GSGSSSKPFV VSARNATQAA NAWRTALTFQ QIKQVEEFCY QPMAVLGYER VNSPEEVKDL SKT Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.

N-acetylglucosamine-6-O-sulfotransferases, such as CHST2, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHST2
Gene Accession No. Q9Y4C5
Gene Alias AI428561; AW121776; C130041E03Rik; C6ST; carbohydrate (chondroitin 6/keratan) sulfotransferase 2; carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2; carbohydrate sulfotransferase 2; Chst2; Chts2; epididymis secretory protein Li 75; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2; GlcNAc6ST; glcNAc6ST-1; GN6ST; Gn6ST-1; GST2; GST-2; HEL-S-75; N-acetylglucosamine 6-O-sulfotransferase 1; N-acetylglucosamine-6-O-sulfotransferase
Gene Symbols CHST2
Host Species Rabbit
Immunogen Recombinant Human CHST2. Recombinant protein control fragment (Product #RP-90506).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9435
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.