Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565472 Supplier Invitrogen™ Supplier No. PA565472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IISRVSDKLKQ IPQALADANS TDPALILAEN ASLLSLSELD SAFSQLQSRL Highest antigen sequence identity to the following orthologs - mouse 84%, rat 86%.

CHST3 is an enzyme which catalyzes the sulfation of chondroitin, a proteoglycan found in the extracellular matrixand most cells which is involved in cell migration and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Chst3
Gene Accession No. Q7LGC8
Gene Alias C6ST; C6ST1; C6ST-1; carbohydrate (chondroitin 6) sulfotransferase 3; carbohydrate (chondroitin 6/keratan) sulfotransferase 3; carbohydrate sulfotransferase 3; chondroitin 6 sulfotransferase 1; chondroitin 6-O-sulfotransferase 1; Chondroitin 6-sulfotransferase; chondroitin 6-sulfotransferase 3; Chst3; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0; GST; Gst0; GST-0; HSD
Gene Symbols Chst3
Host Species Rabbit
Immunogen Recombinant Human CHST3. Recombinant protein control fragment (Product #RP-100930).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9469
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.