Learn More
Invitrogen™ CHST4 Monoclonal Antibody (4D7)

Description
Peptide Sequence: LMIDSRIVMG QHEQKLKKED QPYYVMQVIC QSQLEIYKTI QSLPKALQER YLLVRYEDLA RAPVAQTSRM YEFVGLEFLP HLQTWVHNIT RGKGMGDHAF.
Specifications
Specifications
| Antigen | CHST4 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4D7 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Chst4 |
| Gene Accession No. | Q8NCG5 |
| Gene Alias | carbohydrate (chondroitin 6/keratan) sulfotransferase 4; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4; carbohydrate sulfotransferase 4; Chst4; galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3; galactose/N-acetylglucosamine/N-acetylglucosamine; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3; GlcNAc6ST2; GlcNAc6ST-2; Gn6st-2; Gst3; GST-3; HECGLCNAC6ST; HEC-GlcNAc6ST; high endothelial cell GlcNAC-6-sulphotransferase; high endothelial cells N-acetylglucosamine 6-O-sulfotransferase; L-selectin ligand sulfotransferase; LSST; N-acetylglucosamine 6-O-sulfotransferase 2; sulfo6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.