Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHST4 Mouse anti-Human, Clone: 1E1A4, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 666231IG150UL

Description
This gene encodes an N-acetylglucosamine 6-O sulfotransferase. The encoded enzyme transfers sulfate from 3′phosphoadenosine 5′phospho-sulfate to the 6-hydroxyl group of N-acetylglucosamine on glycoproteins. This protein is localized to the Golgi and is involved in the modification of glycan structures on ligands of the lymphocyte homing receptor L-selectin. Alternate splicing in the 5′ UTR results in multiple transcript variants that encode the same protein.Specifications
CHST4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NCG5 | |
CHST4 | |
CHST4 Fusion Protein Ag3987 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1E1A4 | |
Unconjugated | |
CHST4 | |
CHST4, GlcNAc6ST 2, Gn6st 2, GST 3, HEC GlcNAc6ST, LSST | |
Mouse | |
Protein G | |
RUO | |
10164 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction