Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103701 Supplier Invitrogen™ Supplier No. PA5103701
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CHST5.

The protein encoded by this gene is an enzyme that catalyzes the transfer of a sulfate group to the GlcNAc residues of keratan. Keratan sulfate helps maintain corneal transparency. Defects in this gene are a cause of macular corneal dystrophy (MCD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHST5
Gene Accession No. Q9GZS9
Gene Alias AI173964; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5; carbohydrate sulfotransferase 5; C-GlcNAc6ST; CHST5; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-alpha; GlcNAc6ST-3; Gn6st-3; GST4; GST-4; GST4-alpha; GST4-beta; hIGn6ST; I-GlcNAc6ST; I-GlcNAc-6-ST; intestinal GlcNAc-6-sulfotransferase; intestinal N-acetylglucosamine-6-O-sulfotransferase; MCDC1; mIGn6ST; N-acetylglucosamine 6-O-sulfotransferase 3
Gene Symbols CHST5
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHST5(Accession Q9GZS9), corresponding to amino acid residues A107-C157.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23563
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.