Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHST5/CHST6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529117
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an enzyme that catalyzes the transfer of a sulfate group to the GlcNAc residues of keratan. Keratan sulfate helps maintain corneal transparency. Defects in this gene are a cause of macular corneal dystrophy (MCD).
Specifications
CHST5/CHST6 | |
Polyclonal | |
Unconjugated | |
CHST6 | |
carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6; carbohydrate sulfotransferase 5; Carbohydrate sulfotransferase 6; C-GlcNAc6ST; CHST5; CHST6; corneal N-acetylglucosamine 6-sulfotransferase; corneal N-acetylglucosamine-6-O-sulfotransferase; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-alpha; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta; GlcNAc6ST-3; glcNAc6ST-5; gn6st-3; gn6st-5; GST4-alpha; GST4-beta; hCGn6ST; hIGn6ST; I-GlcNAc6ST; I-GlcNAc-6-ST; intestinal GlcNAc-6-sulfotransferase; Intestinal N-acetylglucosamine-6-O-sulfotransferase; MCDC1; N-acetylglucosamine 6-O-sulfotransferase 3; N-acetylglucosamine 6-O-sulfotransferase 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23563, 4166 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9GZS9, Q9GZX3 | |
CHST5, CHST6 | |
Recombinant fragment corresponding to a region within amino acids 57 and 340 of Human CHST5/6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction