Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116306
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116306 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116306 Supplier Invitrogen™ Supplier No. PA5116306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHST5 Polyclonal Antibody for Western Blot

The protein encoded by this gene is an enzyme that catalyzes the transfer of a sulfate group to the GlcNAc residues of keratan. Keratan sulfate helps maintain corneal transparency. Defects in this gene are a cause of macular corneal dystrophy (MCD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CHST5
Gene Accession No. Q9GZS9
Gene Alias AI173964; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5; carbohydrate sulfotransferase 5; C-GlcNAc6ST; CHST5; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-alpha; GlcNAc6ST-3; Gn6st-3; GST4; GST-4; GST4-alpha; GST4-beta; hIGn6ST; I-GlcNAc6ST; I-GlcNAc-6-ST; intestinal GlcNAc-6-sulfotransferase; intestinal N-acetylglucosamine-6-O-sulfotransferase; MCDC1; mIGn6ST; N-acetylglucosamine 6-O-sulfotransferase 3
Gene Symbols CHST5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 275-302 amino acids from the C-terminal region of human CHST5.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23563
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.