Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHST5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139973
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YRLVRFEDLA REPLAEIRAL YAFTGLTLTP QLEAWIHNIT HGS.
The protein encoded by this gene is an enzyme that catalyzes the transfer of a sulfate group to the GlcNAc residues of keratan. Keratan sulfate helps maintain corneal transparency. Defects in this gene are a cause of macular corneal dystrophy (MCD).
Specifications
CHST5 | |
Polyclonal | |
Unconjugated | |
CHST5 | |
AI173964; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5; carbohydrate sulfotransferase 5; C-GlcNAc6ST; CHST5; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-alpha; GlcNAc6ST-3; Gn6st-3; GST4; GST-4; GST4-alpha; GST4-beta; hIGn6ST; I-GlcNAc6ST; I-GlcNAc-6-ST; intestinal GlcNAc-6-sulfotransferase; intestinal N-acetylglucosamine-6-O-sulfotransferase; MCDC1; mIGn6ST; N-acetylglucosamine 6-O-sulfotransferase 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23563 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9GZS9 | |
CHST5 | |
Recombinant protein corresponding to Human CHST5. Recombinant protein control fragment (Product #RP-109148). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction