Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109542 Supplier Invitrogen™ Supplier No. PA5109542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RIVHLVRDPR AVLRSREQTA KALARDNGIV LGTNGTWVEA DPGLRVVREV CRSHVRIAEA ATLKPPPFLR GRYRLVRFED LAREPLAEIR ALYAFTGLSL TPQLEAWIHN ITHGSGPGAR REAFKTSSRN ALNVSQAWRH ALPFAKIRRV QELCAGALQL LGYRPVYSED EQRNLALDLV LPRGLNGFTW ASSTASHPRN.

Sulfotransferase that utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyze the transfer of sulfate to position 6 of non-reducing N-acetylglucosamine (GlcNAc) residues of keratan. Mediates sulfation of keratan in cornea. Keratan sulfate plays a central role in maintaining corneal transparency. Acts on the non-reducing terminal GlcNAc of short and long carbohydrate substrates that have poly-N- acetyllactosamine structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHST6
Gene Accession No. Q9GZX3
Gene Alias carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6; carbohydrate sulfotransferase 5; carbohydrate sulfotransferase 6; carbohydrate sulfotransferase 6; C-GlcNAc6ST; CHST6; corneal N-acetylglucosamine 6-sulfotransferase; corneal N-acetylglucosamine-6-O-sulfotransferase; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta; glcNAc6ST-5; gn6st-5; GST4-beta; hCGn6ST; LOC489707; MCDC1; N-acetylglucosamine 6-O-sulfotransferase 5
Gene Symbols CHST6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 196-395 of human CHST6 (NP_067628.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4166
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.