Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHST6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548359
Description
Sulfotransferase that utilizes 3′-phospho-5′-adenylyl sulfate (PAPS) as sulfonate donor to catalyze the transfer of sulfate to position 6 of non-reducing N-acetylglucosamine (GlcNAc) residues of keratan. Mediates sulfation of keratan in cornea. Keratan sulfate plays a central role in maintaining corneal transparency. Acts on the non-reducing terminal GlcNAc of short and long carbohydrate substrates that have poly-N- acetyllactosamine structures.
Specifications
CHST6 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9GZX3 | |
CHST6 | |
KLH conjugated synthetic peptide between 296-325 amino acids from the C-terminal region of human CHST6. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CHST6 | |
Carbohydrate sulfotransferase 6, 282-, Corneal N-acetylglucosamine-6-O-sulfotransferase, C-GlcNAc6ST, hCGn6ST, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta, GST4-beta, N-acetylglucosamine 6-O-sulfotransferase 5, GlcNAc6ST-5, Gn6st-5, CHST6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4166 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction