Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHST7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552023
Description
Immunogen sequence: PMWHLWQALY PGDAESLQGA LRDMLRSLFR CDFSVLRLYA PPGDPAARAP DTANLTTAAL FRWRTNKVIC SPPLCPGAPR ARAEVGLVED TACERSCPPV AIRALEAECR KYPVVVIKDV RLLDLGVLVP LLRDPGLNLK VVQL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
This gene belongs to the sulfotransferase gene family. Sulfotransferases generate sulfated glycosaminoglycan (GAG) moities during chondroitin sulfate biosynthesis. They create considerable structural diversity among chondroitin sulfates by transferring sulfate with remarkable specificity for the underlying oligosaccharide substrate. This gene product mainly transfers sulfate to N-acetylgalactosamine. The regulated expression of each member of this gene family may be an important determinant of sulfated GAGs expression and the associated function of chondroitin sulfates as regulators of many biologic processes. This gene is part of a gene cluster on chromosome Xp11.23.
Specifications
CHST7 | |
Polyclonal | |
Unconjugated | |
CHST7 | |
2600013M07Rik; C6ST-2; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 7; carbohydrate (N-acetylglucosamino) sulfotransferase 7; carbohydrate sulfotransferase 7; Chondroitin 6-sulfotransferase 2; chondroitin 6-sulfotransferase 7; chondroitin 6-sulfotransferase-2; CHST7; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 5; glcNAc6ST-4; Gn6st-4; Gst5; GST-5; mC6ST-2; N-acetylglucosamine 6-O-sulfotransferase 1; N-acetylglucosamine 6-O-sulfotransferase 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56548 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NS84 | |
CHST7 | |
Recombinant protein corresponding to Human CHST7. Recombinant protein control fragment (Product #RP-92335). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction