Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHST9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552828 Supplier Invitrogen™ Supplier No. PA552828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGRVEKRREQ KVTSGWGPVK YLRPVPRIMS TEKIQEHITN QNPKFHMPED VREKKENLLL NSERSTRLLT KTSHSQGGDQ ALSKSTGSPT EKLIEKRQGA KTVFNKFSNM NWPVDIHPLN KSLVKDNKWK KTE Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 45%.

Sulfate groups in carbohydrates confer highly specific functions on glycoproteins, glycolipids, and proteoglycans and are critical for cell-cell interaction, signal transduction, and embryonic development. Sulfotransferases, such as CHST9, carry out sulfation of carbohydrates (Hiraoka et al., 2001 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHST9
Gene Accession No. Q7L1S5
Gene Alias 5430438D01Rik; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9; carbohydrate sulfotransferase 9; CHST9; GalNAc-4-O-sulfotransferase 2; GalNAc4ST2; galNAc-4-ST2; GalNAc4ST-2; GalNAc-4-sulfotransferase 2; N-acetylgalactosamine 4-O-sulfotransferase 2; N-acetylgalactosamine-4-O-sulfotransferase 2; UNQ2549/PRO6175
Gene Symbols CHST9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHST9. Recombinant protein control fragment (Product #RP-90373).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83539
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.