Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHSY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550008
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550008 Supplier Invitrogen™ Supplier No. PA550008
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CHSY1 protein.

CHSY1 synthesizes chondroitin sulfate, a glycosaminoglycan expressed on the surface of most cells and in extracellular matrices. Glycosaminoglycan chains are covalently linked to a wide range of core protein families and regulate many biologic processes, including cell proliferation and recognition, extracellular matrix deposition, and morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHSY1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHSY1
Gene Accession No. Q86X52
Gene Alias carbohydrate (chondroitin) synthase 1; carbohydrate synthase 1; chondroitin glucuronyltransferase 1; chondroitin glucuronyltransferase II; chondroitin sulfate synthase 1; Chondroitin synthase 1; CHSY; CHSY1; ChSy-1; chys1; CSS1; glucuronosyl-N-acetylgalactos; glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase 1; KIAA0990; mKIAA0990; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 1; N-acetylgalactosaminyltransferase 1; N-acetylgalactosaminyltransferase II; TPBS; UNQ756/PRO1487; wu:fc27h05; wu:fc27h06; zgc:64191
Gene Symbols CHSY1
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human CHSY1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22856
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.