Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHSY1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548601
Description
CHSY1 synthesizes chondroitin sulfate, a glycosaminoglycan expressed on the surface of most cells and in extracellular matrices. Glycosaminoglycan chains are covalently linked to a wide range of core protein families and regulate many biologic processes, including cell proliferation and recognition, extracellular matrix deposition, and morphogenesis.
Specifications
CHSY1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q86X52, Q6ZQ11 | |
CHSY1 | |
KLH conjugated synthetic peptide between 504-533 amino acids from the Central region of human CHSY1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
CHSY1 | |
Chondroitin sulfate synthase 1, Chondroitin glucuronyltransferase 1, Chondroitin synthase 1, ChSy-1, Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase 1, N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 1, N-acetylgalactosaminyltransferase 1, CHSY1, CHSY, CSS1, KIAA0990 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22856, 269941 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction