Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChT1 antigen Antibody (CT1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128358
Description
ChT1 antigen Monoclonal specifically detects ChT1 antigen in Avian, Chicken samples. It is validated for Flow Cytometry, Immunoprecipitation, In vitro assay.Specifications
ChT1 antigen | |
Monoclonal | |
0.1 mg/mL | |
Flow Cytometry <= 0.2 ug/10^6 cells, Immunoprecipitation 1:10-1:500, In vitro assay | |
Mouse | |
Protein A or G purified | |
RUO | |
Chicken/Quail ChT1 | |
Store at 4C. Do not freeze. | |
IgG1 κ |
Flow Cytometry, Immunoprecipitation, In vitro Assay | |
CT1 | |
PE | |
ChT1 thymocyte antigen, V set and immunoglobulin domain containing, VSIG1 | |
Chicken thymocytes | |
0.1 mg | |
Primary | |
Avian, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction