Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChT1 Polyclonal Antibody, Huabio

Catalog No. 502544551
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4551 Supplier HUABIO RESEARCH INC Supplier No. HA500314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ChT1 Polyclonal antibody specifically detects ChT1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ChT1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene VSIG1
Gene Accession No. Q86XK7, Q9D2J4
Gene Alias 1700062D20Rik; 4930405J24Rik; cell surface A33 antigen; CHT1; ChT1 thymocyte antigen; dJ889N15.1; Glycoprotein A34; GPA34; MGC44287; V-set and immunoglobulin domain containing 1; V-set and immunoglobulin domain-containing protein 1; Vsig1; VSIG1 protein
Host Species Rabbit
Immunogen Synthetic peptide within human ChT1 aa 301-350.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340547, 78789
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.