Learn More
Invitrogen™ CHTF18 Polyclonal Antibody

Description
Immunogen sequence: LLLDILAPKLR PVSTQLYSTR EKQQLASLVG TMLAYSLTYR QERTPDGQYI YRLEPNVEEL CRFPELPARK PLTYQTKQL Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
Specifications
Specifications
| Antigen | CHTF18 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Chtf18 |
| Gene Accession No. | Q8WVB6 |
| Gene Alias | 6030457M03Rik; C16orf41; C321D2.2; C321D2.3; C321D2.3 (novel protein); C321D2.4; C321D2.4 (novel protein); Chl12; Chl12 homolog; chromosome transmission fidelity factor 18; chromosome transmission fidelity protein 18 homolog; chromosome transmission fidelity protein 18 homolog; LOW QUALITY PROTEIN: chromosome transmission fidelity protein 18 homolog; Chtf18; Ctf18; CTF18, chromosome transmission fidelity factor 18; CTF18, chromosome transmission fidelity factor 18 homolog; CTF18, chromosome transmission fidelity factor 18 homolog (S. cerevisiae); hCTF18; homolog of yeast CHL12; RUVBL; some homology with holliday junction DNA helicase RUVB like; zgc:113153 |
| Gene Symbols | Chtf18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.