Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHTF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566778 Supplier Invitrogen™ Supplier No. PA566778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGPGSGPNLRA GVLLTSGNGP PNPRPVGLGP GPNPNLRSGF LGTNPAPRSG VFPGPG Highest antigen sequence identity to the following orthologs - mouse 84%, rat 47%.

Potential tumor suppressor. Inhibits prostate tumor cell growth, when overexpressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHTF8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHTF8
Gene Accession No. P0CG13
Gene Alias chromosome transmission fidelity factor 8; chromosome transmission fidelity protein 8 homolog; CHTF8; CTF8; CTF8, chromosome transmission fidelity factor 8 homolog; decreased expression in renal and prostate cancer protein; DERPC; hCTF8
Gene Symbols CHTF8
Host Species Rabbit
Immunogen Recombinant Human CHTF8. Recombinant protein control fragment (Product #RP-107436).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54921
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.