Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chums™ 3mm Universal Fit Nylon Rope Eyewear Retainers
Shop All Chums Ltd Products
Click to view available options
Product Type:
Universal-fit Retainer
Color:
Assorted
Red
Description
Patented Universal Fit tips are molded directly to the cord and will accommodate a wide variety of eyeglass frames
- Adjustable slider bead for secure, reliable fit
- Field tested eyewear retainer
- Logo option available
- Made in the USA
Specifications
Specifications
| Color | Assorted |
| Product Type | Universal-fit Retainer |
| Includes | Adjustable slider bead |
| Material | Nylon 3 mm perlon |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction