Learn More
Chums™ Fire Resistant Eyewear Retainer
Catalog No. p-4099380 Shop All Chums Ltd Products
Encompass
Click to view available options
Product Type:
Eyewear retainer, Adjustable, No tail
Catalog No. 19-808-7039
Supplier: Chums™ 12405
Only null left
Add to Cart
Add to Cart
Will not ignite, smolder, melt or drip when exposed to arc flash, direct flame, sparks or welding spatter
  • PBI/Kevlar material
  • NFPA 70E Certified
  • Cut and puncture resistant
  • Adjustable non-conductive PVC slider bead
  • Integrity and suppleness retained up to 540C, LOI = 41-38%
  • Fits most safety eyewear
  • Made in the USA
Compliance
NFPA 70E
Specifications
Product Type Eyewear retainer, Adjustable, No tail
For Use With (Equipment) Fits most safety eyewear
Includes Adjustable bead
Material PBI, Dupont™ Kevlar™ aramid fiber
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.