Learn More
Chums™ Fire Resistant Eyewear Retainer
Shop All Chums Ltd Products
Click to view available options
Product Type:
Eyewear retainer, Adjustable, No tail
Description
- PBI/Kevlar material
- NFPA 70E Certified
- Cut and puncture resistant
- Adjustable non-conductive PVC slider bead
- Integrity and suppleness retained up to 540C, LOI = 41-38%
- Fits most safety eyewear
- Made in the USA
Compliance
NFPA 70E
Specifications
Specifications
| Product Type | Eyewear retainer, Adjustable, No tail |
| For Use With (Equipment) | Fits most safety eyewear |
| Includes | Adjustable bead |
| Material | PBI, Dupont™ Kevlar™ aramid fiber |