Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHURC1 Monoclonal Antibody (2F9)
GREENER_CHOICE

Catalog No. PIMA524502
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524502 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524502 Supplier Invitrogen™ Supplier No. MA524502
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MCGDCVGKEY PNRGNTCLEN GSFLLNFTGC AVCSKRDFML ITNKSLKEED GEEIVTYDHL CKNCHHVIAR HEYTFSIMDE FQEYTMLCLL CGKAEDTISI LPDDPRQMTL LF.

Transcriptional activator that mediates FGF signaling during neural development. Plays a role in the regulation of cell movement. Does not bind DNA by itself.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHURC1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F9
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CHURC1
Gene Accession No. Q8WUH1
Gene Alias 1700030P15Rik; C14orf52; CHCH; CHURC1; churchill domain containing 1; My015; Protein Churchill
Gene Symbols CHURC1
Host Species Mouse
Immunogen CHURC1 (AAH20550, 1 a.a. ~ 112 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91612
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.