Learn More
chx 10 (Visual system homeobox 2) (CT) Sheep anti-Chicken, Human, Mouse, Rat, Polyclonal, Exalpha

Sheep Polyclonal Antibody
Supplier: Vector Laboratories X1179P

Description
This antibody detects Keratin 5 and 14 by Western blot at 58 kDa, corresponding to Keratin 5, and 50 kDa, corresponding to Keratin 14. Does not work on paraffin embedded tissue sections.
chx 10 (Visual system homeobox 2) (CT) Polyclonal antibody specifically detects chx 10 (Visual system homeobox 2) (CT) in Chicken, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Western BlotSpecifications
chx 10 (Visual system homeobox 2) (CT) | |
Polyclonal | |
Unconjugated | |
VSX2 | |
C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2 | |
Antibody raised against a recombinant protein corresponding to amino acids 264 to 361 derived from the C terminal of the human Chx10 protein conjugated to KLH. | |
250 μg | |
Primary | |
Chicken, Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Frozen), Western Blot | |
0.5-2 mg/mL | |
PBS with 0.08% sodium azide; pH | |
P58304, Q61412, Q9IAL1 | |
Sheep | |
Ammonium sulfate precipitation | |
RUO | |
12677, 171360, 338917, 395536 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.