Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chx 10 (Visual system homeobox 2) (NT) Sheep anti-Chicken, Human, Mouse, Rat, Polyclonal, Exalpha
SDP

Catalog No. 502101941
Encompass
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1941 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1941 Supplier Vector Laboratories Supplier No. X1180P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

This antibody detects Keratin 5 & 8 by Western blot at 58 kDa, corresponding to Keratin 5, and 52.5 kDa, corresponding to Keratin 8.

chx 10 (Visual system homeobox 2) (NT) Polyclonal antibody specifically detects chx 10 (Visual system homeobox 2) (NT) in Chicken, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Western Blot

Specifications

Antigen chx 10 (Visual system homeobox 2) (NT)
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.5-2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide; pH
Gene VSX2
Gene Accession No. P58304, Q61412, Q9IAL1
Gene Alias C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2
Host Species Sheep
Immunogen Recombinant protein corresponding to amino acids 1 to 131 derived from the N terminal of the human Chx10 protein conjugated to KLH.
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12677, 171360, 338917, 395536
Target Species Chicken, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.