Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHX10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582092
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582092 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582092 Supplier Invitrogen™ Supplier No. PA582092
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGLNKEPPSS HPRAALDGLA PGHLLAARSV LSPAGVGGMG LLGPGGLPGF YTQPTFLEVL SDPQSVHLQP LGRASGPLDT SQTASSDSED VSSSDRKMSK SALNQTKKRK KRRHRTIFTS YQLEELEKAF NEAH Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

CHX10 encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHX10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VSX2
Gene Accession No. P58304
Gene Alias C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2
Gene Symbols VSX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VSX2. Recombinant protein control fragment (Product #RP-89128).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171360, 338917
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.