Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHX10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674794
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-794 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-794 Supplier Invitrogen™ Supplier No. PA112565
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

By Western blot, this antibody detects a ~46 kDa band. A suggested positive control for this product is rat or mouse retinal tissue lysate.

CHX10 encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHX10
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene VSX2
Gene Accession No. P58304, Q61412, Q9IAL1
Gene Alias C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2
Gene Symbols VSX2
Host Species Sheep
Immunogen Protein corresponding to residues 264-361 from the C terminus of human Chx10, conjugated to KLH.
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12677, 171360, 338917, 395536
Target Species Human, Mouse, Rat, Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.