Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHX10 Recombinant Rabbit Monoclonal Antibody (4Z4R2)
GREENER_CHOICE

Catalog No. PIMA538025
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538025 Supplier Invitrogen™ Supplier No. MA538025
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse spinal cord, Mouse brain, Rat spinal cord, Rat brain Immunogen sequence: AAAESGRKPE GERQALPKLD KMEQDERGPD AQAAISQEEL RENSIAVLRA KAQEHSTKVL GTVSGPDSLA RSTEKPEEEE AMDEDRPAER LSPPQLEDMA.

CHX10 encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHX10
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4Z4R2
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide, pH 7.3
Gene VSX2
Gene Accession No. Q61412
Gene Alias C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2
Gene Symbols VSX2
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12677, 171360
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.