Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHX10 Sheep anti-Human, Mouse, Rat, Chicken, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody
$243.50 - $487.50
Specifications
Antigen | CHX10 |
---|---|
Dilution | Western Blot 0.5 - 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Frozen |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CHX10 Polyclonal antibody specifically detects CHX10 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
CHX10 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Sheep | |
Vision | |
PBS | |
338917 | |
IgG | |
Ammonium sulfate precipitation |
Western Blot 0.5 - 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Frozen | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Chicken | |
ceh-10 homeo domain containing homolog, ceh-10 homeo domain containing homolog (C. elegans), ceh-10 homeodomain containing homolog (C. elegans), Ceh-10 homeodomain-containing homolog, CHX10MCOPCB3, Homeobox protein CHX10, HOX10C elegans ceh-10 homeo domain-containing homolog, MCOP2, RET1, visual system homeobox 2 | |
Antibody raised in sheep against a recombinant protein corresponding to amino acids 1 to 131 derived from the N-terminal region of the human CHX10 protein conjugated to KLH. (Uniprot: P58304) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title