Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chymase/CMA1/Mast Cell Chymase Rabbit anti-Human, Mouse, Rat, Clone: 2O4P8, Novus Biologicals™
SDP

Catalog No. NB165875 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165875 20 μg
NB165874 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165875 Supplier Novus Biologicals Supplier No. NBP31538920UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Chymase/CMA1/Mast Cell Chymase Monoclonal antibody specifically detects Chymase/CMA1/Mast Cell Chymase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Chymase/CMA1/Mast Cell Chymase
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2O4P8
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCT1, MGC119890, MGC119891
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 148-247 of human Chymase/CMA1/Mast Cell Chymase (CMA1) (P23946). GWGRTGVLKPGSDTLQEVKLRLMDPQACSHFRDFDHNLQLCVGNPRKTKSAFKGDSGGPLLCAGVAQGIVSYGRSDAKPPAVFTRISHYRPWINQILQAN
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology, Innate Immunity, Mast Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1215
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.