Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chymotrypsin-like protease Antibody (SR1373), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Chymotrypsin-like protease |
---|---|
Clone | SR1373 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Chymotrypsin-like protease Monoclonal antibody specifically detects Chymotrypsin-like protease in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Chymotrypsin-like protease | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
chymotrypsin-like, chymotrypsin-like protease CTRL-1, CTRL1, EC 3.4.21, EC 3.4.21.-, MGC70821 | |
A synthesized peptide derived from human Chymotrypsin-like protease (Uniprot #: P40313) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1373 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1506 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title