Learn More
Invitrogen™ Chymotrypsin Recombinant Rabbit Monoclonal Antibody (102)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA530445
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CTRC/chymotrypsin C.
This gene encodes a member of the peptidase S1 family. The encoded protein is a serum calcium-decreasing factor that has chymotrypsin-like protease activity. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
Specifications
Chymotrypsin | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q99895 | |
CTRC | |
Recombinant Human CTRC/chymotrypsin C protein (Met1-Leu268). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
102 | |
Unconjugated | |
CTRC | |
1810044E12Rik; caldecrin; chymotrypsin C; chymotrypsin C (caldecrin); chymotrypsin-C; CLCR; Ctrc; ELA4; elastase 4; elastase IV; preprocaldecrin; serum calcium decreasing factor; serum calcium-decreasing factor | |
Rabbit | |
Protein A | |
RUO | |
11330 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.