Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CI 994, Tocris Bioscience™
SDP

Catalog No. 295210 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-5210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-5210 Supplier R&D Systems Supplier No. 2952/10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Class I histone deacetylase inhibitor; orally active

Orally active class I histone deacetylase (HDAC) inhibitor (Ki values are 0.41, 0.75, >100 and >100 μM for HDAC1, HDAC3, HDAC6 and HDAC8 respectively). Mediates G1 cell cycle arrest, inhibits proliferation and induces apoptosis in vitro and in vivo. Increases neuroplasticity during memory extinction. Also increases efficiency of hepatic differentiation of hPSCs

Chemical Identifiers

CAS 112522-64-2
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
MDL Number MFCD00866266
InChI Key VAZAPHZUAVEOMC-UHFFFAOYSA-N
Synonym tacedinaline, acetyldinaline, 4-acetamido-n-2-aminophenyl benzamide, 4-acetylamino-n-2'-aminophenyl benzamide, tacedinalina, ci994 tacedinaline, n-acetyldinaline, tacedinaline usan:inn, tacedinalina inn-spanish, 4-acetylamino-n-2-aminophenyl benzamide
PubChem CID 2746
IUPAC Name N-(2-aminophenyl)-4-acetamidobenzamide
SMILES CC(=O)NC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1N

Specifications

Quantity 10 mg
Formula Weight 269.3
Percent Purity >99%
Chemical Name or Material CI 994
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.