Learn More
Siemens Healthineers Ci-Trol™ Heparin Controls
Catalog No. p-7235898 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-044-652 Low
23-044-653 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-652 Supplier Siemens Healthineers Supplier No. 10445715
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Controls for heparin assay procedures.
Ci-Trol™ heparin low and high controls are used used to monitor the performance of heparin therapy using the activated partial thromboplastin time (APTT).
TRUSTED_SUSTAINABILITY
Specifications
Type Herapin Control
Level Low
Quantity 10 x 1 mL