Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIAO1 Antibody (6C9), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242631B
Description
CIAO1 Monoclonal specifically detects CIAO1 in Human samples. It is validated for Western Blot, ELISA.Specifications
CIAO1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CIAO1 | |
Recombinant human CIAO1 (1-339aa) purified from E. coli | |
100 μL | |
Cell Cycle and Replication | |
9391 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
6C9 | |
Western Blot, ELISA | |
CIA1, cytosolic iron-sulfur protein assembly 1, cytosolic iron-sulfur protein assembly 1 homolog (S. cerevisiae), probable cytosolic iron-sulfur protein assembly protein CIAO1, WD repeat domain 39, WD repeat-containing protein 39, WD40 protein Ciao1, WDR39cytosolic iron-sulfur protein assembly 1 homolog | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction