Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cIAP-2/HIAP-1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | cIAP-2/HIAP-1 |
---|---|
Concentration | 1.5 mg/mL |
Dilution | Western Blot 1 ug/ml, Simple Western 1:100, Immunohistochemistry 1:800-1:3200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:800-1:3200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP127972SS
![]() |
Novus Biologicals
NBP127972SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP127972
![]() |
Novus Biologicals
NBP127972 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
cIAP-2/HIAP-1 Polyclonal specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
cIAP-2/HIAP-1 | |
Western Blot 1 ug/ml, Simple Western 1:100, Immunohistochemistry 1:800-1:3200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:800-1:3200 | |
Polyclonal | |
Rabbit | |
Apoptosis, Inhibitors Activators and Regulators | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49 | |
BIRC3 | |
IgG | |
Affinity Purified | |
64 kDa |
1.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q13489 | |
330 | |
Synthetic peptide made to an internal portion of human cIAP2 (within residues 100-200). [Swiss-Prot# Q13489] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title