Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ cIAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520066
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520066 Supplier Invitrogen™ Supplier No. PA520066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects both c-IAP1 and c-IAP2. A suggested positive control is human lung tissue lysate. PA5-20066 can be used with blocking peptide PEP-0185.

The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. This encoded protein inhibits apoptosis induced by serum deprivation and menadione, a potent inducer of free radicals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cIAP1
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene BIRC2
Gene Accession No. Q13490, Q62210
Gene Alias Api1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW146227; baculoviral IAP repeat containing 2; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 2; baculoviral IAP repeat-containing protein 2; putative inhibitor of apoptosis; BIRC2; Birc3; cellular inhibitor of apoptosis 1; cIAP1; C-IAP1; cIAP2; HIAP1; HIAP2; Hiap-2; IAP homolog B; IAP1; IAP2; IAP-2; inhibitor of apoptosis protein; inhibitor of apoptosis protein 2; LOC100622859; mcIAP1; MIAP1; MIAP2; mIAP-2; MIHB; MIHC; NFR2-TRAF signalling complex protein; rIAP1; RING finger protein 48; RING-type E3 ubiquitin transferase BIRC2; RNF48; TNFR2-TRAF-signaling complex protein 2
Gene Symbols BIRC2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids at the C-terminus of human c-IAP1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11797, 329
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.