Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ cIAP2 Monoclonal Antibody (OTI3G4)
GREENER_CHOICE

Catalog No. PIMA526360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526360 Supplier Invitrogen™ Supplier No. MA526360
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

cIAP2 Monoclonal Antibody for Western Blot, ICC/IF, Flow

The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. The encoded protein inhibits apoptosis induced by serum deprivation but does not affect apoptosis resulting from exposure to menadione, a potent inducer of free radicals. The amino acid sequence predicts three baculovirus IAP repeat domains and a ring finger domain. Transcript variants encoding the same isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cIAP2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BIRC3
Gene Accession No. Q13489
Gene Alias AIP 2; AIP1; Api1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW107670; baculoviral IAP repeat containing 3; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 3; Birc2; Birc3; Cellular inhibitor of apoptosis 2; cIAP1; cIAP-1; cIAP2; C-IAP2; cIAP-2; H HIAP1; H MALT2; HAIP1; HIAP1; hIAP-1; HIAP2; IAP homolog; IAP homolog C; IAP1; IAP-1; IAP2; Inhibitor of apoptosis protein 1; MALT2; mammalian IAP homolog C; MIAP1; mIAP-1; MIAP2; MIHB; MIHC; RING finger protein 49; RING-type E3 ubiquitin transferase BIRC3; RNF49; TNFR2-TRAF signaling complex protein; TNFR2-TRAF-signaling complex protein 1
Gene Symbols BIRC3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 255-529 of BIRC3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 330
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.